2-(Diphenylphosphino)benzenesulfonic acid 98%

Supplier: Ambeed
Warning

A641184-1G A641184-250MG
77267-056EA 60 USD
77267-056 77267-058
2-(Diphenylphosphino)benzenesulfonic acid 98%
2-(Diphenylphosphino)benzenesulfonic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₅O₃PS
MDL Number: MFCD20921518
CAS Number: 111864-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR