1-Bromoheptane 95%

Supplier: Ambeed
Danger

Synonyms: 1-Heptyl bromide

A641154-10G A641154-25G
77242-972EA 5.15 USD
77242-972 77242-974
1-Bromoheptane 95%
1-Bromoheptane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₅CH₃
MW: 179.1 g/mol
MDL Number: MFCD00000273
CAS Number: 629-04-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR