(R)-2-(1-Aminoethyl)-4-chlorophenol 98%

Supplier: Ambeed
Warning

A640883-250MG A640883-1G
77242-970EA 10.3 USD
77242-970 77747-320
(R)-2-(1-Aminoethyl)-4-chlorophenol 98%
(R)-2-(1-Aminoethyl)-4-chlorophenol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD06762061
CAS Number: 1228571-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR