3,4-Dimethoxybenzyl chloride 95%

Supplier: Ambeed
Danger

A640252-100MG A640252-1G A640252-250MG A640252-5G
77280-468EA 6 USD
77280-468 77280-470 77280-472 77280-474
3,4-Dimethoxybenzyl chloride 95%
3,4-Dimethoxybenzyl chloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MDL Number: MFCD00185554
CAS Number: 7306-46-9
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR