5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid, (5-(Dimethylcarbamoyl)-2-fluorophenyl)boronic acid, 5-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

A639746-1G A639746-250MG A639746-5G
77254-594EA 106.09 USD
77254-594 77254-596 77254-598
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 98%
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 211.00 g/mol
MDL Number: MFCD08235102
CAS Number: 874289-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR