Dimethyl-3,3'-thiodipropionate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 3,3-Thiodipropionate, 3,3'-Thiodipropionic Acid Dimethyl Ester

A639508-1000G A639508-25G A639508-500G
77280-460EA 97.85 USD
77280-460 77280-462 77280-464
Dimethyl-3,3'-thiodipropionate 98%
Dimethyl-3,3'-thiodipropionate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄S
MW: 206.26 g/mol
MDL Number: MFCD00008465
CAS Number: 4131-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR