Sodium 4-aminohippurate 98%

Supplier: Ambeed
Warning

Synonyms: Aminohippurate sodium, Sodium (4-aminobenzoyl)glycinate

A639297-100MG A639297-1MG A639297-25MG
77280-454EA 8.24 USD
77280-454 77280-456 77280-458
Sodium 4-aminohippurate 98%
Sodium 4-aminohippurate

Precaución: for Research Use Only

Formula: C₉H₉N₂NaO₃
MW: 216.16 g/mol
MDL Number: MFCD00044689
CAS Number: 94-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR