2-Bromo-4,6-difluorophenol 98%

Supplier: Ambeed
Warning

A639154-1G A639154-25G A639154-5G
77292-968EA 13 USD
77292-968 77292-970 77292-972
2-Bromo-4,6-difluorophenol 98%
2-Bromo-4,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MDL Number: MFCD00070752
CAS Number: 98130-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR