(15aR)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine 98%

Supplier: Ambeed
Warning

A638945-1G A638945-250MG
77267-046EA 721 USD
77267-046 77267-048
(15aR)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine 98%
(15aR)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine

Precaución: for Research Use Only

Formula: C₄₁H₃₆O₂P₂
CAS Number: 301847-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR