5-(3-(4-Methoxyphenyl)allylidene)pyrimidine-2,4,6(1H,3H,5H)-trione 98%

Supplier: Ambeed
Warning

A638881-10MG A638881-1MG A638881-25MG A638881-5MG
77242-962EA 40 USD
77242-962 77242-964 77242-966 77242-968
5-(3-(4-Methoxyphenyl)allylidene)pyrimidine-2,4,6(1H,3H,5H)-trione 98%
5-(3-(4-Methoxyphenyl)allylidene)pyrimidine-2,4,6(1H,3H,5H)-trione

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD01177921
CAS Number: 100872-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR