2-Chloro-4-fluorocinnamic acid 98% (only trans isomers)

Supplier: Ambeed
Warning

A638747-100MG A638747-1G A638747-25G A638747-5G
77267-038EA 5.15 USD
77267-038 77267-040 77267-042 77267-044
2-Chloro-4-fluorocinnamic acid 98% (only trans isomers)
2-Chloro-4-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
MDL Number: MFCD00236027
CAS Number: 133220-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR