α-Bromo-2-chlorophenylacetic acid 97%

Supplier: Ambeed
Warning

A638540-100G A638540-10G A638540-25G A638540-5G
77242-954EA 105 USD
77242-954 77242-956 77242-958 77242-960
α-Bromo-2-chlorophenylacetic acid 97%
α-Bromo-2-chlorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD03428513
CAS Number: 29270-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR