(S)-5-benzyl-2-mesityl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%

Supplier: Ambeed
Danger

A638221-100MG A638221-1G A638221-250MG A638221-50MG A638221-5G
77254-572EA 45 USD
77254-572 77254-574 77254-576 77254-578 77254-580
(S)-5-benzyl-2-mesityl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%
(S)-5-benzyl-2-mesityl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₄BF₄N₃O
CAS Number: 950842-73-6
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR