Benzo[1,2-b:6,5-b']dithiophene-4,5-dione 97%

Supplier: Ambeed
Warning

A638115-100MG A638115-1G A638115-250MG A638115-5G
77267-028EA 67.98 USD
77267-028 77267-030 77267-032 77267-034
Benzo[1,2-b:6,5-b']dithiophene-4,5-dione 97%
Benzo[1,2-b:6,5-b']dithiophene-4,5-dione

Precaución: for Research Use Only

Formula: C₁₀H₄O₂S₂
MW: 220.26 g/mol
MDL Number: MFCD25372694
CAS Number: 24243-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR