Pyrimidine-2-carboximidamide 95%

Supplier: Ambeed
Warning

A637571-1G A637571-250MG
77280-450EA 195.7 USD
77280-450 77280-452
Pyrimidine-2-carboximidamide 95%
Pyrimidine-2-carboximidamide

Precaución: for Research Use Only

Formula: C₅H₆N₄
MW: 122.13 g/mol
MDL Number: MFCD09755980
CAS Number: 45695-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR