6'-(Ethyl(isopentyl)amino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 97%

Supplier: Ambeed
Warning

Synonyms: HUN16415

A637217-10G A637217-1G A637217-5G A637217-100G A637217-25G
77242-944EA 28 USD
77242-944 77242-946 77242-948 77617-688 77617-690
6'-(Ethyl(isopentyl)amino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 97%
6'-(Ethyl(isopentyl)amino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

Precaución: for Research Use Only

Formula: C₃₄H₃₄N₂O₃
MDL Number: MFCD11044531
CAS Number: 70516-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR