2,6-Dichlorobenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dichlorobenzene-1-sulphonyl chloride

A637043-10G A637043-1G A637043-5G
77292-962EA 20.6 USD
77292-962 77292-964 77292-966
2,6-Dichlorobenzenesulfonyl chloride 95%
2,6-Dichlorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
MDL Number: MFCD00052311
CAS Number: 6579-54-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR