tert-Butyl (5-ethynylpyridin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A636909-100MG A636909-1G A636909-250MG
77242-938EA 37 USD
77242-938 77242-940 77242-942
tert-Butyl (5-ethynylpyridin-3-yl)carbamate 97%
tert-Butyl (5-ethynylpyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD13189734
CAS Number: 1192472-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR