2,3-Dihydro-1,4-benzodioxin-6-ylmethanol 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-6-(hydroxymethyl)-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanol

A636552-1G A636552-250MG A636552-5G
77254-566EA 39.14 USD
77254-566 77254-568 77254-570
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol 95%
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD01317581
CAS Number: 39270-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR