(9H-Fluoren-9-yl)methyl (2-hydroxyethyl)(methyl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A636413-5G
A636413-25G
77242-936EA
14
USD
77242-936
77747-311
(9H-Fluoren-9-yl)methyl (2-hydroxyethyl)(methyl)carbamate 98%
(9H-Fluoren-9-yl)methyl (2-hydroxyethyl)(methyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₈H₁₉NO₃ MW: 297.34 g/mol |
MDL Number:
MFCD03788643 CAS Number: 147687-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...