2-Fluoro-5-iodobenzotrifluoride 97%

Supplier: Ambeed
Warning

A636317-10G A636317-5G A636317-100G
77267-016EA 8 USD
77267-016 77267-018 77749-301
2-Fluoro-5-iodobenzotrifluoride 97%
2-Fluoro-5-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄I
MDL Number: MFCD00236678
CAS Number: 59382-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR