3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-7-hydroxy-4-methylcoumarin

A636299-10G A636299-1G A636299-5G
77280-444EA 299 USD
77280-444 77280-446 77280-448
3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one 97%
3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₃
MDL Number: MFCD00047643
CAS Number: 86-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR