1-Benzyl-3-cyclohexyl-1-(piperidin-4-ylmethyl)urea 98%

Supplier: Ambeed
Warning

A636073-100MG A636073-10MG A636073-1MG A636073-25MG A636073-50MG A636073-5MG
77267-004EA 232 USD
77267-004 77267-006 77267-008 77267-010 77267-012 77267-014
1-Benzyl-3-cyclohexyl-1-(piperidin-4-ylmethyl)urea 98%
1-Benzyl-3-cyclohexyl-1-(piperidin-4-ylmethyl)urea

Precaución: for Research Use Only

Formula: C₂₀H₃₁N₃O
MDL Number: MFCD01475985
CAS Number: 1629138-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR