(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,5S)-(+)-1-(2-(1,3-Dioxolan-2-yl)phenyl-2,5-dimethylphospholane, 2-{2-[(2S,5S)-2,5-Dimethylphospholan-1-yl]phenyl}-1,3-dioxolane, (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
A635961-100MG
A635961-1G
A635961-250MG
A635961-5G
77254-558EA
66.95
USD
77254-558
77254-560
77254-562
77254-564
(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane ≥98%, ee 99%
(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Precaución: for Research Use Only
Formula:
C₁₅H₂₁O₂P MW: 264.305 g/mol |
MDL Number:
MFCD09265097 CAS Number: 695816-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...