(4-Ethoxy-2-fluorophenyl)methanol 97%

Supplier: Ambeed
Warning

A635464-100MG A635464-1G A635464-250MG A635464-5G
77242-928EA 32 USD
77242-928 77242-930 77242-932 77242-934
(4-Ethoxy-2-fluorophenyl)methanol 97%
(4-Ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MDL Number: MFCD28519144
CAS Number: 1393470-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR