2-(9H-Carbazol-9-yl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(9H-Carbazol-9-yl)benzeneboronic acid, (2-(9H-Carbazol-9-yl)phenyl)boronic acid

A635296-10G A635296-1G A635296-5G A635296-25G
77254-546EA 82 USD
77254-546 77254-548 77254-550 77551-922
2-(9H-Carbazol-9-yl)phenylboronic acid 95%
2-(9H-Carbazol-9-yl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₄BNO₂
MDL Number: MFCD30543408
CAS Number: 1189047-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR