2-Chloro-5-cyanophenylboronic acid 98%

Supplier: Ambeed
Warning

A635164-1G A635164-250MG A635164-5G
77280-432EA 32.96 USD
77280-432 77280-434 77280-436
2-Chloro-5-cyanophenylboronic acid 98%
2-Chloro-5-cyanophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClNO₂
MW: 181.3841 g/mol
MDL Number: MFCD06656271
CAS Number: 936249-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR