Ethyl 4-amino-3-cyanobenzoate 97%

Supplier: Ambeed
Warning

A634937-100MG A634937-1G A634937-250MG
77242-918EA 13 USD
77242-918 77242-920 77242-922
Ethyl 4-amino-3-cyanobenzoate 97%
Ethyl 4-amino-3-cyanobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD16038777
CAS Number: 1260742-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR