4-Acetamido-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A634752-1G A634752-25G A634752-5G
77266-986EA 9 USD
77266-986 77266-988 77266-990
4-Acetamido-3-nitrobenzoic acid 98%
4-Acetamido-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₅
MW: 224.17
MDL Number: MFCD00014703
CAS Number: 1539-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR