3-(Difluoromethoxy)benzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Borono-3-(difluoromethoxy)benzene, (3-(Difluoromethoxy)phenyl)boronic acid

A634457-100MG A634457-1G A634457-250MG A634457-5G
77280-424EA 7 USD
77280-424 77280-426 77280-428 77280-430
3-(Difluoromethoxy)benzeneboronic acid 97%
3-(Difluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
MDL Number: MFCD08706282
CAS Number: 866607-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR