6-Chloro-5-methyl-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A633486-100MG A633486-250MG A633486-50MG
77242-898EA 128.75 USD
77242-898 77242-900 77242-902
6-Chloro-5-methyl-1H-benzo[d][1,2,3]triazole 95%
6-Chloro-5-methyl-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.59 g/mol
MDL Number: MFCD00238834
CAS Number: 221343-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR