2,5-Dimethyl-3(2H)-furanone 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dimethylfuran-3(2H)-one

A633485-1G A633485-25G A633485-5G
77254-536EA 6.18 USD
77254-536 77254-538 77254-540
2,5-Dimethyl-3(2H)-furanone 98%
2,5-Dimethyl-3(2H)-furanone

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00052571
CAS Number: 14400-67-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR