BAY 87-2243 98%

Supplier: Ambeed
Warning

Synonyms: 5-(1-((2-(4-cyclopropylpiperazin-1-yl)pyridin-4-yl)methyl)-5-methyl-1H-pyrazol-3-yl)-3-(4-(trifluoromethoxy)phenyl)-1,2,4-oxadiazole

A633112-10MG A633112-1MG A633112-25MG A633112-50MG A633112-5MG A633112-100MG
77242-888EA 46.35 USD
77242-888 77242-890 77242-892 77242-894 77242-896 77617-674
BAY 87-2243 98%
BAY 87-2243

Precaución: for Research Use Only

Formula: C₂₆H₂₆F₃N₇O₂
MW: 525.53 g/mol
MDL Number: MFCD28143915
CAS Number: 1227158-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR