5-[(3,5-Difluorophenyl)methyl]-1H-indazol-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-(3,5-difluorobenzyl)-1H-indazole

A633080-1G A633080-250MG A633080-5G
77254-528EA 49.44 USD
77254-528 77254-530 77747-302
5-[(3,5-Difluorophenyl)methyl]-1H-indazol-3-amine 95%
5-[(3,5-Difluorophenyl)methyl]-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₂N₃
MW: 259.254 g/mol
MDL Number: MFCD28129098
CAS Number: 1108745-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR