2,3-Dimethoxybenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2,3-Dimethoxyphenyl)methanol

A632794-10G A632794-1G A632794-25G A632794-5G
77254-520EA 76.22 USD
77254-520 77254-522 77254-524 77254-526
2,3-Dimethoxybenzyl alcohol 95%
2,3-Dimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00004612
CAS Number: 5653-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR