(S)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A632107-100MG A632107-1G A632107-250MG A632107-5G
77254-512EA 64 USD
77254-512 77254-514 77254-516 77254-518
(S)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl 98%
(S)-2,2'-Bis(diphenylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₄₀H₃₆O₄P₂
MDL Number: MFCD19443622
CAS Number: 1365531-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR