3-(Propylamino)propane-1,2-diol 97%

Supplier: Ambeed
Danger

A631978-100MG A631978-1G A631978-250MG
77254-506EA 66.95 USD
77254-506 77254-508 77254-510
3-(Propylamino)propane-1,2-diol 97%
3-(Propylamino)propane-1,2-diol

Precaución: for Research Use Only

Formula: C₆H₁₅NO₂
MW: 133.18 g/mol
MDL Number: MFCD11213268
CAS Number: 70189-88-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR