9,10-Bis(pyridin-4-ylethynyl)anthracene 97%

Supplier: Ambeed
Warning

A631391-100MG A631391-250MG A631391-5G
77266-968EA 50.47 USD
77266-968 77266-970 77266-972
9,10-Bis(pyridin-4-ylethynyl)anthracene 97%
9,10-Bis(pyridin-4-ylethynyl)anthracene

Precaución: for Research Use Only

Formula: C₂₈H₁₆N₂
MW: 380.44 g/mol
CAS Number: 596130-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR