(4-Tolyloxy)acetic acid 98%

Supplier: Ambeed
Warning

A631383-100G A631383-1G A631383-25G A631383-5G
77254-498EA 132 USD
77254-498 77254-500 77254-502 77254-504
(4-Tolyloxy)acetic acid 98%
(4-Tolyloxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(CH₃))OCH₂COOH
MDL Number: MFCD00014365
CAS Number: 940-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR