2-Chloro-4-(methylsulphonyl)benzoic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

A630748-25G A630748-5G A630748-100G A630748-500G
77242-882EA 16 USD
77242-882 77242-884 77617-662 77747-300
2-Chloro-4-(methylsulphonyl)benzoic acid 98%
2-Chloro-4-(methylsulphonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
MDL Number: MFCD00216496
CAS Number: 53250-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR