1-Bromo-2,3,5-trifluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2,3,5-Trifluorobromobenzene

A630286-1G A630286-25G A630286-5G
77292-944EA 5.15 USD
77292-944 77292-946 77292-948
1-Bromo-2,3,5-trifluorobenzene 98%
1-Bromo-2,3,5-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
MDL Number: MFCD00012232
CAS Number: 133739-70-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR