Tris(4-nonylphenyl) phosphite 97%

Supplier: Ambeed
Danger

A630245-10G A630245-25G
77266-964EA 7.21 USD
77266-964 77266-966
Tris(4-nonylphenyl) phosphite 97%
Tris(4-nonylphenyl) phosphite

Precaución: for Research Use Only

Formula: C₄₅H₆₉O₃P
MW: 689.00 g/mol
MDL Number: MFCD00015301
CAS Number: 3050-88-2
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR