4-(tert-Butyl)-1H-pyrazol-3-amine 97%
Supplier: Ambeed
|
Warning
|
A630004-100MG
A630004-1G
A630004-250MG
77242-876EA
152.44
USD
77242-876
77242-878
77242-880
4-(tert-Butyl)-1H-pyrazol-3-amine 97%
4-(tert-Butyl)-1H-pyrazol-3-amine
Precaución: for Research Use Only
Formula:
C₇H₁₃N₃ MW: 139.19 g/mol |
MDL Number:
MFCD27995843 CAS Number: 255047-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...