3-Hydroxy-1-naphthoic acid 97%

Supplier: Ambeed
Warning

A629530-100MG A629530-1G A629530-250MG
77254-486EA 27 USD
77254-486 77594-932 77594-934
3-Hydroxy-1-naphthoic acid 97%
3-Hydroxy-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₃
MW: 188.17 g/mol
MDL Number: MFCD17011843
CAS Number: 19700-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR