N-(5-(4-Acetylpiperazin-1-yl)pyridin-2-yl)-2-(2'-fluoro-3-methyl-[2,4'-bipyridin]-5-yl)acetamide 98+%

Supplier: Ambeed
Warning

A629360-10MG A629360-1MG A629360-25MG A629360-5MG
77242-868EA 78 USD
77242-868 77242-870 77242-872 77242-874
N-(5-(4-Acetylpiperazin-1-yl)pyridin-2-yl)-2-(2'-fluoro-3-methyl-[2,4'-bipyridin]-5-yl)acetamide 98+%
N-(5-(4-Acetylpiperazin-1-yl)pyridin-2-yl)-2-(2'-fluoro-3-methyl-[2,4'-bipyridin]-5-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₄H₂₅FN₆O₂
MDL Number: MFCD30343851
CAS Number: 1243245-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR