3-((1,3-Dioxoisoindolin-2-yl)methyl)benzoic acid 95%

Supplier: Ambeed
Warning

A629286-100MG A629286-1G A629286-250MG
77280-402EA 52 USD
77280-402 77280-404 77280-406
3-((1,3-Dioxoisoindolin-2-yl)methyl)benzoic acid 95%
3-((1,3-Dioxoisoindolin-2-yl)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₁NO₄
MDL Number: MFCD00418210
CAS Number: 106352-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR