2-Methylstyrene 95% stabilized

Supplier: Ambeed
Danger

Synonyms: 2-Vinyltoluene, 1-Methyl-2-vinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A629272-1G A629272-5G
77280-398EA 23.69 USD
77280-398 77280-400
2-Methylstyrene 95% stabilized
2-Methylstyrene

Precaución: for Research Use Only

Formula: C₉H₁₀
MW: 118.18 g/mol
MDL Number: MFCD00014936
CAS Number: 611-15-4
UN: 2618
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR