2,2,2-trifluoroethylhydrazine 65% in water

Supplier: Ambeed
Danger

Synonyms: (2,2,2-Trifluoroethyl)hydrazine, 2,2,2-Trifluoroethylhydrazine

A629193-10G A629193-1G A629193-25G
77266-958EA 36 USD
77266-958 77266-960 77617-658
2,2,2-trifluoroethylhydrazine 65% in water
2,2,2-trifluoroethylhydrazine

Precaución: for Research Use Only

Formula: C₂H₅F₃N₂
MDL Number: MFCD00007622
CAS Number: 5042-30-8
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR