3-(Chlorosulfonyl)-4-methoxybenzoic acid 95%

Supplier: Ambeed
Danger

A629000-1G A629000-250MG
77280-394EA 60 USD
77280-394 77280-396
3-(Chlorosulfonyl)-4-methoxybenzoic acid 95%
3-(Chlorosulfonyl)-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₅S
MW: 250.65 g/mol
MDL Number: MFCD00024880
CAS Number: 50803-29-7
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR