Pamapimod 98+%

Supplier: Ambeed
Warning

Synonyms: Ro-4402257, Ro 4402257, R1503, R-1503, Pamapimod, Ro4402257, 6-(2,4-Difluorophenoxy)-2-((1,5-dihydroxypentan-3-yl)amino)-8-methylpyrido[2,3-d]pyrimidin-7(8H)-one, R 1503

A628708-10MG A628708-1MG A628708-25MG A628708-5MG
77266-944EA 57.68 USD
77266-944 77266-946 77266-948 77266-950
Pamapimod 98+%
Pamapimod

Precaución: for Research Use Only

Formula: C₁₉H₂₀F₂N₄O₄
MW: 406.38 g/mol
MDL Number: MFCD25976551
CAS Number: 449811-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR